StabilZyme® AP Conjugate stabilizer, Surmodics

Supplier: Surmodics

SA01-1000
102466-272EA 379.2 CAD
102466-272
StabilZyme® AP Conjugate stabilizer, Surmodics
Protein Linking and Labeling Systems

StabilZyme® AP and StabilZyme® Protein-Free AP stabilizers provide assay developers a complete set of tools to stabilize different AP conjugates for long term, in-solution stability.


  • Extended shelf life of kits
  • Increased signal-to-noise ratios
  • Eliminates the need for subsequent diluents


StabilZyme® AP Stabilizer specifically optimizes the AP enzyme and is the gold standard for in-solution conjugate stability.


Certifications: ISO 13485:2016 & 9001:2015 Certified

Order Now

SPECIFICATIONS

Environmentally Preferable
pH 5.5-6.5
Host
Protein BSA
Storage buffer MES
Storage temperature 2 to 8° C or Room Temperature
Shelf life 2.5 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR