Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Supplier: Labconco
112816002 112826002 112836002 112016002 112026002 112036002 112116002 112126002 112136002 112216002 112226002 112236002
10221-900EA 41001 CAD
10221-900 10221-902 10221-904 10221-906 10222-154 10222-156 10222-158 10222-160 10222-162 10222-164 10222-166 10222-168
Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Fume Hoods and Enclosures Chemical Fume Hoods
Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are suitable for general chemistry applications and have the dimensions required for over-sized equipment.
These hoods are constructed of durable, powder-coated steel with four tempered safety glass sashes and a corrosion- and chemical-resistant, fiberglass-reinforced composite panel liner that surpasses all national codes for flame spread. Features include removable access panels, 73.5" loading height and pre-wired LED lighting rated for 50000 hours.
Laboratory workers with applications requiring distillationMore Product Information