BRAND® BLAUBRAND® Graduated Cylinders, Tall Form, Class A, USP, Certified, BrandTech® Scientific
Supplier: Brandtech
32805 32808 32820 32828 32838 32848 32854 32862 32864
10218-734PK 95.01 CAD
10218-734 10218-736 10218-738 10218-740 10218-742 10218-744 10218-746 10218-748 10218-750
BRAND® BLAUBRAND® Graduated Cylinders, Tall Form, Class A, USP, Certified, BrandTech® Scientific
Cylinders Measuring Cylinders
BRAND® BLAUBRAND® tall form graduated cylinders are certified Class A and constructed to DIN EN ISO standards of Boro 3.3 glass and meet the error limits of USP/ASTM.
  • Individually adjusted
  • Flexible stencils for accuracy at partial volumes
  • Durable, highly legible BLAUBRAND® ink (blue line) for easy adjustment and maximum precision
Flexible stencils are used for maximum accuracy of all intermediate volumes. High quality inks are baked on during an annealing process that relieves thermal stresses. This provides volume stability even after heating up to 25More Product Information