BLAUBRAND® Bulb Pipettes, 1 Mark, Class AS, USP, Certified, BrandTech
Supplier: Brandtech
30602 30603 30605 30606 30607 30608 30609 30610 30611 30612 30613 30614 30615 30616 30617 30618 30619 30601
10218-650PK 260.33 CAD
10218-650 10218-652 10218-654 10218-656 10218-658 10218-660 10218-662 10218-664 10218-666 10218-668 10218-670 10218-672 10218-674 10218-676 10218-678 10218-680 10218-682 76313-890
BLAUBRAND® Bulb Pipettes, 1 Mark, Class AS, USP, Certified, BrandTech
Pipettes Graduated Pipettes
Class AS volumetric single mark pipettes.
  • Certified Class A
  • Calibrated to deliver (TD)
  • Constructed according to DIN EN ISO standards with error limits that meet the specifications of USP/ASTM
  • Each individual piece is marked on a computer controlled, automated production line to ensure maximum precision
  • USP batch certificates are supplied
  • Volumetrically stable even when heated up to 250 °C via special annealing process
  • Durable, high-contrast blue enamel markings
BLAUBRAND® single mark bulb pipettes are individually marked to provide Class A 'to deliver'More Product Information