Zone Samplers, Bürkle
Supplier: BURKLE INC
5316-0055 5316-0085 5316-0150 5316-1055 5316-1085 5316-1150 5316-2055 5316-2085 5316-2150 5316-3055 5316-3150 5317-1058 5317-1085 5317-1150 5317-2055 5317-2085 5317-2150 5319-1085 5319-1150 5319-1250 5319-2085 5319-2150 5319-2250
10217-068EA 1670.74 CAD
10217-068 10217-070 10217-072 10217-074 10217-076 10217-078 10217-080 10217-082 10217-084 10217-086 10217-088 10217-090 10217-096 10217-098 10217-100 10217-102 10217-104 10217-106 10217-108 10217-110 10217-112 10217-114 10217-116 10217-118
Zone Samplers, Bürkle
Samplers Solid Samplers
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain.
  • Easy to clean, removable tip
  • Colour coding (Multi and Uno samplers)
  • Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
  • 3 lengths from 550 to 1500 mm, Jumbo up to 2500 mm
  • Samplers have smallest possible gap between inner and outer tubes
Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno samplerMore Product Information