Heated Incubators, 7002 Series, Caron Products
Supplier: Caron Products
7002-10-1 7002-10-2 7002-10-3 7002-25-1 7002-25-2 7002-25-3 7002-33-1 7002-33-2 7002-33-3
10216-764EA 32605.13 CAD
10216-764 10216-766 10216-768 10216-770 10216-772 10216-774 10216-776 10216-778 10216-780
Heated Incubators, 7002 Series, Caron Products
Incubators Standard Incubators
Caron’s Heated Incubators offer controlled temperature conditions (10°C above ambient to 70°C)
115V units will plug into a standard laboratory electrical outlet, contributing to cost savings and energy efficiency. Temperature uniformity is excellent for reliable, repeatable results. Key standard features include: touchscreen user interface; built-in preventative maintenance; tool-less removable interior; sliding shelves; horizontal airflow system that generates gentle and evenly distributed airflow across all shelf locations; heated, triple pane glass door (708L and 934 L models); andMore Product Information