Tough-Tag Thermal Transfer Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77591-2 77591-3 77591-4 77591-5 77599-10 77599-20 77599-30 77599-40 77599-50
102098-636EA 234.77 CAD
102098-636 102098-638 102098-640 102098-642 102098-742 102098-744 102098-746 102098-748 102098-750
Tough-Tag Thermal Transfer Labels, Electron Microscopy Sciences
Labels
Thermal Transfer Tough-Tags are a pre-cut, peel-off label designed for use in thermal transfer printers. They are a reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles and other laboratory containers.
  • Resists degradation from scuffing, chemicals, moisture and wide temperature fluctuations
  • Print as little as one label at a time, eliminating label waste and saving time and money
  • Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Specially coated for use 
More Product Information