Opaque White, Extra Fine Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72168-01
102097-878EA 20.46 CAD
102097-878
Opaque White, Extra Fine Pen, Electron Microscopy Sciences
Pens Permanent Markers
Permanently mark on nearly any surface in white color and the marking is waterproof
This pen is ideal for SEM, marking sample identification on carbon tabs, aluminum stubs, conductive tape or any similar surface. The pen contains xylene.