Reagent Reservoirs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70495-10 70495-20 70495-30 70495-35 70495-40 70496-10 70496-20 70496-30 70496-35 70497-10 70497-20 70497-30 70497-35 70497-40
102096-962CS 308.98 CAD
102096-962 102096-964 102096-966 102096-968 102096-970 102096-972 102096-974 102096-976 102096-978 102096-982 102096-984 102096-986 102096-988 102096-990
Reagent Reservoirs, Electron Microscopy Sciences
Reagent Reservoirs
Reagent reservoirs are disposable and available in both sterile and non sterile varieties.
  • Unique "trough within a trough" maximizes reagent recovery
  • Reservoir has Pour-off spouts on all four corners
  • Wide base provides stability and avoids spills
  • 25 ml Reservoir has a divider accommodates eight channel pipets on one side and one to four channel pipets on the other
  • Offered in bulk cases or individually wrapped (ideal for use in clean rooms)
The Sidecar Reservoir features “Two-in-One design” for precise reagent adding/mixing and “Sidecar” holds asMore Product Information