Autoradiography Ruler, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68550-04
102096-324EA 91.34 CAD
102096-324
Autoradiography Ruler, Electron Microscopy Sciences
Rulers
The Autoradiography Ruler Set (ARS) is an orientation and measurement device that exposes metric ruler markings directly onto X-ray film.
  • Phosphorescent, non-radioactive metric rulers for radiographs
  • Make easy electrophoretic mobility calculations
Utilizes a specially formulated phosphorescent dye compound and a unique emission filter which serves as an ideal non-radioactive substitute for ink markings. The phosphorescent emission of each ruler can be regenerated indefinitely and can be reused any number of times. Produces a high resolution image thatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable