Fuchs-Resenthal Count Chamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63512-10
102094-782EA 1342.59 CAD
102094-782
Fuchs-Resenthal Count Chamber, Electron Microscopy Sciences
Slides Counting Chambers
Designed for counting leukocytes, eosinophils and cells in cerebrospinal fluid.
Grid pattern is etched directly onto the glass surface. The chamber is 0.2 mm deep. Chamber set includes: one chamber and 2 octagonal cover glasses.