Dumont Tweezers Style 3, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0109-3-PO
102091-528EA 101.75 CAD
102091-528 102091-558
Dumont Tweezers Style 3, Electron Microscopy Sciences
Forceps
Thin, 120 mm tips.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400° C (DIN 50 914) and is suitable for autoclave sterilization at 270°C. This Dumont patented alloy is composedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable