Parafilm M, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70991 70990
100501-338EA 107.28 CAD
100501-338 102091-164
Parafilm M, Electron Microscopy Sciences
Seals General Purpose Sealing Films
Laboratory film is used to prevent evaporation, contamination and hydration. Seals labware quickly and efficiently. It is ideal for electron microscopy drop staining and to pick up thin films when coating grids.