Anti-SERCA2 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1720
10209-206EA 1442.03 CAD
10209-206
Anti-SERCA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Sarcoplasmic/endoplasmic reticulum calcium ATPase 2(ATP2A2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: SERCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR