Anti-IgM Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Boster Biological Technology
BA1116-0.25
10208-958EA 439.75 CAD
10208-958
Anti-IgM Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR