Anti-IgM Antibody (HRP (Horseradish Peroxidase))

Supplier: Boster Biological Technology
BA1075-0.25
10208-934EA 439.75 CAD
10208-934
Anti-IgM Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR