Anti-DR5 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1842
10208-820EA 1442.03 CAD
10208-820
Anti-DR5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 10B(TNFRSF10B) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: DR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol DR5
Antigen name Death Receptor 5
Environmentally Preferable
Antigen synonyms TRAIL receptor 2|TRAILR2|CD262 antigen|CD262|ZTNFR9.|Death domain containing receptor for TRAIL/Apo2L|TRICK2|KILLER|TRICK2A|TRICKB|Death receptor 5|p53 regulated DNA damage inducible cell death receptor (killer)|TNFRSF10B|CD 262|KILLER/DR5|DR 5|TRICK2B|TNF related apoptosis inducing ligand receptor 2|Fas like protein precursor|TRAIL R2|Tumor necrosis factor receptor like protein ZTNFR9|TRANCER|Tumor necrosis factor receptor superfamily member 10b|TNF-related apoptosis-inducing ligand receptor 2|DR5
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Epitope near the C-terminus
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human DR5 (333-348aa DPTETLRQCFDDFADL).
Amino acid number 333 to 348
Sequence DPTETLRQCFDDFADL
Form Lyophilized
Purification purified by immunogen affinity purification
Cross adsorption No
Storage temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR