Human IL-22 PicoKine ELISA Kit, Boster
Supplier: Boster Biological Technology
EK0933
10207-852EA 1940.4 CAD
10207-852
Human IL-22 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Human IL-22
Ordering information: Kits include 96-well plate precoated with anti- human IL-22 antibody 1; ST0000-10 Lyophilized recombinant human IL-22 standard 10ng/tubex2; AR1107 Biotinylated anti- human IL-22 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.