Anti-IL-6 Polyclonal Antibody

Supplier: Boster Biological Technology
PB9034
10207-514EA 1441.96 CAD
10207-514
Anti-IL-6 Polyclonal Antibody
Antibodies
Polyclonal antibody for IL 6/IL6 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Mouse. IL 6/IL6 information: Molecular Weight: 24384 MW; Subcellular Localization: Secreted.

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR