Anti-STAT4 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1692
10206-904EA 1442.03 CAD
10206-904
Anti-STAT4 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 4(STAT4) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR