Anti-STAT1 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1508
10206-838EA 1442.03 CAD
10206-838
Anti-STAT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 1-alpha/beta(STAT1) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR