Anti-CYP11A1 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1698-1
10206-642EA 1442.03 CAD
10206-642
Anti-CYP11A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Cholesterol side-chain cleavage enzyme, mitochondrial(CYP11A1) detection. Tested with WB in Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: CYP11A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR