Anti-CASP3 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1849
10206-506EA 1440.57 CAD
10206-506
Anti-CASP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Caspase-3(CASP3) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR