Rat sL-Selectin PicoKine ELISA Kit, Boster

Supplier: Boster Biological Technology

EK0937
10205-496EA 1940.24 CAD
10205-496
Rat sL-Selectin PicoKine ELISA Kit, Boster
Assays ELISAs

Sandwich ELISA kit of Quantitative Detection for Rat sL-Selectin


Ordering information: Kits include 96-well plate precoated with anti- rat sL-Selectin antibody 1; ST0000-10 Lyophilized recombinant rat sL-Selectin standard 40ng/tubex2; AR1107 Biotinylated anti- rat sL-Selectin antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.

Order Now

SPECIFICATIONS

Target protein sL-selectin
Environmentally Preferable
Primary antibody reactivity Rat
Assay Type Sandwich
Host
Immunogen Standard: NSO, W39-N332

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR