EDU Culture Dish Control System, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
04200504212 0420050421 04200504211
10199-852KT 5522.45 CAD
10199-852 10199-854 10199-856
EDU Culture Dish Control System, Bioptechs Inc.®
Educational Classroom Kits and Activities
The EDU Culture Dish Control System is a simplified version of the Delta T® system
  • Enhance the instruction of biology
  • Includes curriculum relevant demonstrations
  • Temperature selection options
  • Basic X, Y translation built in
The EDU Starter Set includes all of the necessary components required for full system operation. The EDU Controller and Power Supply is an electronic, closed-loop temperature controller interfaced to EDU stage adapters.
It is designed to facilitate demonstrations of live cellular events in a classroom setting.
Certifications:More Product Information