Stable "Z"® Starter Set, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
4031926
10199-838KT 7760.34 CAD
10199-838
Stable "Z"® Starter Set, Bioptechs Inc.®
Heaters Hot Plates
The Stable "Z"® is a device designed to warm biological specimens in standard plasticware on a microscope stage, with the distinct advantage of eliminating "Z" axis drift.
  • Minimal mass of heating component
  • Specimen is coplanar with support surface to eliminate "Z" axis drift
  • Minimizes heat transfer to the stage
  • Has option of heated lid to prevent condensation
  • Adjustable specimen plane to accommodate various dishes
  • Option for Nunc Lab-Tec multi-well slides
  • Includes a reference temperature probe
Certifications: CE certified.
Ordering information: SetMore Product Information