Pratt Uterine Dilators, OR Grade, Sklar
Supplier: Sklar
90-5215 90-5219 90-5223 90-5227 90-5231 90-5235 90-5239 90-5243 90-5250
10194-422EA 275.12 CAD
10194-422 10199-272 10199-274 10199-276 10199-278 10199-280 10199-282 10199-284 10199-286
Pratt Uterine Dilators, OR Grade, Sklar
Dilators
Sklar's Pratt Uterine Dilator are a double-ended OB/GYN instrument used for progressive dilation of the uterus.
  • Available in several sizes
  • Dilator has two different size tips
  • Straight in the middle, slightly curved at each end, with a smooth, rounded tip
These are most frequently used during intra-uterine procedures such as D&C and D&E. The size of these dilators is determined by the circumference of the tip and measured in French units, with one French unit equalling 1/3 mm.
In comparison to other intra-uterine dilators, Pratt dilatorsMore Product Information