Sklar Black™ Steiner-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3560 90-3561 90-3562
10197-200EA 2158.36 CAD
10197-200 10197-202 10197-204
Sklar Black™ Steiner-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Black™ Steiner-Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in laser OB/GYN procedures such as vaginal hysterectomy or dilation and curettage.
  • Stainless steel with reduced reflectivity polymer coating
  • This speculum features extra long blades
  • Weighted with a medium, 21/2 lbs.
Features a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable