Adson Retractor, OR Grade, Sklar®
Supplier: Sklar
42-1510 42-1511 42-1611 42-1775 42-1785
10196-680EA 1133.01 CAD
10196-680 10196-682 10196-684 10196-690 10196-692
Adson Retractor, OR Grade, Sklar®
Retractors
Adson Retractor is a finger ring, self-retaining retractor used during intracranial procedures to hold back the cerebellum
It is available with sharp or blunt interlocking prongs in various configurations, angled or straight arms, and multiple lengths.