Cheron Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
90-7910
10196-412EA 490.42 CAD
10196-412
Cheron Dressing Forceps, OR Grade, Sklar
Forceps
Cheron Dressing Forceps are used for holding swabs and dressings in gynecological procedures.
Their long length makes them good for use in IUD insertion and removal procedures. The handles of this instrument are angled sideways so the users hands do not obstruct the view of the surgical site.
Order Now
SPECIFICATIONS
Environmentally Preferable