Bakes Dilators, OR Grade, Sklar®
Supplier: Sklar
32-1530 32-1533 32-1534 32-1535 32-1536 32-1537 32-1538 32-1539 32-1610 32-1611
10195-478EA 1992.39 CAD
10195-478 10195-480 10195-482 10195-484 10195-486 10195-488 10195-658 10195-660 10195-662 10195-664
Bakes Dilators, OR Grade, Sklar®
Dilators
Sklar's® Bakes Dilators are a single-ended instrument with a solid, bullet-shaped tip used to dilate and enlarge a duct.
The Bakes Dilator is most commonly used to open the bile duct to allow bile to pass to the liver. The shaft of this instrument is thin, round, and maleable to improve movement within the duct. It has a standard, overall length of 83/4 to 9" with variable tip sizes.