Sklarlite™ Sklarcut™ Operating Scissors, OR Grade, Sklar®
Supplier: Sklar
23-1260 23-1262 23-1264 23-1266
10195-280EA 413.59 CAD
10195-280 10195-282 10195-284 76192-944
Sklarlite™ Sklarcut™ Operating Scissors, OR Grade, Sklar®
Scissors
Sklarlite™ Sklarcut™ Operating scissors, also known as Surgical scissors, are a versatile surgical instrument
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklarlite™ Sklarcut™ scissors are a lighter version of the standard scissors, that feature micro-serrations on one blade to assist in securely holding the tissue during cutting and minimize tissue from slippingMore Product Information