Rochester-Ochsner Forceps, OR Grade, Sklar®
Supplier: Sklar
17-2690 17-2890 17-2962 18-1752 18-1852 17-2685 17-2885
10194-774EA 321.02 CAD
10194-774 10194-778 10194-782 10194-828 10194-830 76192-222 76192-224
Rochester-Ochsner Forceps, OR Grade, Sklar®
Forceps
Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held
Censitrac Ready OR Grade Rochester-Ochsner Forceps are industry standard.More Product Information