Merit™ Allis Tissue Forceps, Physician Grade, Sklar
Supplier: Sklar
97-930 97-934 97-938
10184-936EA 49.69 CAD
10184-936 10184-938 10194-484
Merit™ Allis Tissue Forceps, Physician Grade, Sklar
Forceps
Merit™ Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue.
They are used with tonsil, vaginal, breast, and thyroid tissue.
Merit™ instruments are office-grade line of products used for routine and in-office procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable