Tischler-Kevorkian Micro-Cut Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8920
10194-148EA 3112.08 CAD
10194-148
Tischler-Kevorkian Micro-Cut Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Tischler-Kevorkian Micro-Cut Biopsy Punch is an OB/GYN instrument used in colposcopy procedures to obtain cervical tissue samples for biopsy.
This product features a single lower tooth and a single upper tooth used to grasp difficult to reach tissue. It also has a locking mechanism above the horn on the handle, so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal.
This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers.
Order Now
SPECIFICATIONS
Environmentally Preferable