Ballenger Sponge Forceps, OR Grade, Sklar®
Supplier: Sklar
74-1470 74-1370 74-1570 74-1670
10192-476EA 216.72 CAD
10192-476 10198-602 10198-604 10198-606
Ballenger Sponge Forceps, OR Grade, Sklar®
Forceps
Ballenger Sponge Forceps are a ratcheted and finger ring instrument used to grasp tissue or hold sponges
Sponge Forceps may also be known as sponge sticks. The instrument is used to hold gauze or cotton, that is wrapped around the tips and the ratcheted is locked to secure a firm grip. This creates a sponge stick that can be used in any type of procedure to absorb blood and excess fluids from the surgical site and surrounding areas.
They may be known as Foerster Sponge Forceps, however the shorter length instrument is often referred to as Ballenger.
Order Now
SPECIFICATIONS
Environmentally Preferable