Merit™ Webster Needle Holder, Physician Grade, Sklar
Supplier: Sklar
97-809
10192-338EA 44.05 CAD
10192-338
Merit™ Webster Needle Holder, Physician Grade, Sklar
Forceps
Merit™ Instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures
These cost effective, high-quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is curved with smooth, blunt tips.