Microliter™ 800 Series Syringes, Hamilton
Supplier: Hamilton
84850 84851 84852 84854 84856 84858 84857 84861 84853 84855 84859 84890 7642-01 7646-01
60374-100EA 333.84 CAD
60374-100 60374-102 60374-122 60374-144 60374-166 60374-188 60374-280 60374-326 60374-246 60374-268 60374-304 89221-470 10192-124 10192-130
Microliter™ 800 Series Syringes, Hamilton
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. The 800 series syringes are a reinforced plunger version of the original 700 series. Attached to the flange of the syringe is a removable blue syringe holder that prevents heat transfer, dispense inaccuracies and plunger blow outs. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL upMore Product Information