Preadipocyte/Adipocyte Medium, PromoCell
Supplier: Promocell
C-27410 C-27411 C-27415 C-27417 C-27431 C-27435 C-27437 C-27438 C-27439
10172-352EA 355.48 CAD
10172-352 10172-354 10172-356 10172-358 10172-360 10172-362 10172-364 10172-366 10172-368 10172-370
Preadipocyte/Adipocyte Medium, PromoCell
Cell Culture Supplements and Additives
The PromoCell Preadipocyte Growth Medium is a low-serum (5% V/V) medium optimized for the expansion of human preadipocytes
Recommended for Human White Preadipocytes (HWP), Human Adipocytes
Ordering information: PromoCell Preadipocyte/Adipocyte Media are available as Medium (Ready-touse) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as SupplementMix and SupplementPacks can also be purchased separately.