Anti-ASS1 Mouse Monoclonal Antibody [clone: 2C10]

Supplier: Genetex
GTX60552
10169-002EA 939.42 CAD
10169-002
Anti-ASS1 Mouse Monoclonal Antibody [clone: 2C10]
Antibodies
Mouse Monoclonal Antibody [2C10] to ASS1

Type: Primary
Antigen: ASS1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR