Anti-CS Mouse Monoclonal Antibody [clone: GT1761]

Supplier: Genetex
GTX628143
10168-034EA 930.11 CAD
10168-034
Anti-CS Mouse Monoclonal Antibody [clone: GT1761]
Antibodies
Mouse Monoclonal antibody to Citrate synthetase (citrate synthase)

Type: Primary
Antigen: CS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR