Anti-USP7 Mouse Monoclonal Antibody [clone: GT6512]

Supplier: Genetex
GTX631109
10167-642EA 930.11 CAD
10167-642
Anti-USP7 Mouse Monoclonal Antibody [clone: GT6512]
Antibodies
Mouse Monoclonal antibody [GT6512] to USP7 (ubiquitin specific peptidase 7 (herpes virus-associated))

Type: Primary
Antigen: USP7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR