Anti-UQCRC1 Mouse Monoclonal Antibody [clone: GT139]

Supplier: Genetex
GTX630413
10167-576EA 930.11 CAD
10167-576
Anti-UQCRC1 Mouse Monoclonal Antibody [clone: GT139]
Antibodies
Mouse Monoclonal antibody [GT139] to UQCRC1 (ubiquinol-cytochrome c reductase core protein I)

Type: Primary
Antigen: UQCRC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR