Anti-CHAC2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX128819
10166-812EA 1026.27 CAD
10166-812
Anti-CHAC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CHAC2 (ChaC, cation transport regulator homolog 2 (E. coli))

Type: Primary
Antigen: CHAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR