Anti-ARHGAP1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX101472
10166-448EA 924.96 CAD
10166-448
Anti-ARHGAP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ARHGAP1 (Rho GTPase activating protein 1)

Type: Primary
Antigen: ARHGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ARHGAP1
Environmentally Preferable
Antigen name Rho GTPase Activating Protein 1
Antigen synonyms RHOGAP1|RHOGAP|p50rhoGAP|CDC42GAP
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 392
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ARHGAP1. The exact sequence is proprietary.
Purification Purified by antigen-affinity chromatography.
Concentration 0.31 mg/ml
Storage buffer 0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
Storage temperature Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR