Anti-RERG Rabbit Polyclonal Antibody

Supplier: Genetex
GTX117011
10166-186EA 930.11 CAD
10166-186
Anti-RERG Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RERG (RAS-like, estrogen-regulated, growth inhibitor)

Type: Primary
Antigen: RERG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR