Anti-PBXIP1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX116669
10166-168EA 930.11 CAD
10166-168
Anti-PBXIP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PBXIP1 (pre-B-cell leukemia homeobox interacting protein 1)

Type: Primary
Antigen: PBXIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR