Anti-RBCK1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX115451
10166-122EA 930.11 CAD
10166-122
Anti-RBCK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RBCK1 (RanBP-type and C3HC4-type zinc finger containing 1)

Type: Primary
Antigen: RBCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR