Anti-RNASEH2A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX115433
10166-114EA 930.11 CAD
10166-114
Anti-RNASEH2A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RNASEH2A (ribonuclease H2, subunit A)

Type: Primary
Antigen: RNASEH2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR