Anti-Sarcosine Rabbit Polyclonal Antibody

Supplier: Genetex
GTX13479
10164-628EA 982.84 CAD
10164-628
Anti-Sarcosine Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Sarcosine

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR